Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Styren ≥99% stabilised

Styren ≥99% stabilised

Glass bottle, 2.500 mL
Katalog # A18481.0F
Sundhed
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
Styren ≥99% stabilised
Katalog # A18481.0F
CAS nummer:  100-42-5
Styren ≥99% stabilised
Styren ≥99% stabilised

Glass bottle, 2.500 mL

CAS nummer:  100-42-5
Price
840,00 kr
Log ind for tilgængelighed
1 * 2.500 mL
Product Details & Documents

Stabilized with 10-15 ppm 4-tert-butylcatechol

Purity: 99% CAS No.: 100-42-5 Molecular Formula: C8H8 Formula Weight: 104.15 Storage Temperature: 2°C to 8°C Flash Point: 31°(88°F) Refractive Index: 1.5460 Odour: Pungent pH: neutral Melting Point: -31° Boiling Point: 145-146° Beilstein Reference: 1071236 Merck Reference: 14,8860 UN No.: 2055 MDL No.: MFCD00008612

Synonymer:  Phenylethene, Phenylethylene, Vinylbenzene
  • CAS nummer:100-42-5
  • Formel:C₈H₈
  • MDL nummer:MFCD00008612
  • Molecular Weight:104.15 g/mol
  • Merck Index:14,8860
  • Kogende Pt:145…146 °C
  • Smeltende Pt:–31 °C
  • Tæthed:0,906
  • Flash Pt:31 °C (88 °F)
  • Opbevaringstemperatur:Køleskab
  • FN:2055
  • ADR:3,III
  •  :stabiliseret med 4-tert-Butylpyrocatechol 10-15 ppm
  • EINECS:202-851-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2.500 mL

Specifikationer