Order Entry
Denmark
Orders LinkContactUsLinkComponent
(R)-(-)-Hydratropasyre ≥97%

(R)-(-)-Hydratropasyre ≥97%

Glass bottle for solids, 1 g
Katalog # A17639.03
Irriterende
Leverandør:   Thermo Fisher Scientific
(R)-(-)-Hydratropasyre ≥97%
Katalog # A17639.03
CAS nummer:  7782-26-5
(R)-(-)-Hydratropasyre ≥97%
(R)-(-)-Hydratropasyre ≥97%

Glass bottle for solids, 1 g

CAS nummer:  7782-26-5
Price
2.360,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Purity: 97% CAS No.: 7782-26-5 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 148°(298°F) Refractive Index: 1.5230 Melting Point: 32-34° Boiling Point: 264-266° Beilstein Reference: 2207688 Optical Rotation: -73° (c=1.6 in chloroform) MDL No.: MFCD00063140

Synonymer:  (R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid
  • CAS nummer:7782-26-5
  • Formel:C₉H₁₀O₂
  • MDL nummer:MFCD00063140
  • Molecular Weight:150.18 g/mol
  • Kogende Pt:264…266 °C
  • Smeltende Pt:32…34 °C
  • Tæthed:1,100
  • Flash Pt:148 °C (298 °F)
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer