Methyl rød natrium salt
Glass bottle for solids, 500 g
CAS No.: 845-10-3 Molecular Formula: C15H14N3NaO2 Formula Weight: 291.29 Beilstein Reference: 4065488 MDL No.: MFCD00002426
: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate
- :845-10-3
- :C₁₅H₁₄N₃NaO₂
- :MFCD00002426
- :291.28 g/mol
- :13,06142
- :> 180 °C
- :Stue temperatur
- :212-682-9
- Pakning:Glass bottle for solids
- Pakn:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



