Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Methyl rød natrium salt

Methyl rød natrium salt

Glass bottle for solids, 100 g
Katalog # A17455.22
Leverandør:   Thermo Fisher Scientific
Methyl rød natrium salt
Katalog # A17455.22
CAS nummer:  845-10-3
Methyl rød natrium salt
Methyl rød natrium salt

Glass bottle for solids, 100 g

CAS nummer:  845-10-3
Price
633,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

CAS No.: 845-10-3 Molecular Formula: C15H14N3NaO2 Formula Weight: 291.29 Beilstein Reference: 4065488 MDL No.: MFCD00002426

Synonymer:  Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate
  • CAS nummer:845-10-3
  • Formel:C₁₅H₁₄N₃NaO₂
  • MDL nummer:MFCD00002426
  • Molecular Weight:291.28 g/mol
  • Merck Index:13,06142
  • Smeltende Pt:> 180 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:212-682-9
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.