Orders LinkContactUsLinkComponent
Bromcresol grønt
Bromcresol grønt
Glass bottle for solids, 5 g
Katalog # A17090.06
Leverandør:   Thermo Fisher Scientific
Bromcresol grønt
Katalog # A17090.06
CAS nummer:  76-60-8
Bromcresol grønt
Bromcresol grønt
Glass bottle for solids, 5 g
CAS nummer:  76-60-8
Price
468,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
CAS No.: 76-60-8 Molecular Formula: C21H14Br4O5S Formula Weight: 698.02 Melting Point: ca 225° dec. Beilstein Reference: 372527 Merck Reference: 14,1386 MDL No.: MFCD00005874
Synonymer:  3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS nummer:76-60-8
  • Formel:C₂₁H₁₄Br₄O₅S
  • MDL nummer:MFCD00005874
  • Molecular Weight:698.02 g/mol
  • Merck Index:14,1386
  • Smeltende Pt:∼ 225 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-972-8
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g