Orders LinkContactUsLinkComponent
Phenolphthalein ≥98%
Phenolphthalein ≥98%
Glass bottle for solids, 250 g
Katalog # A17135.30
Sundhed
Leverandør:   Thermo Fisher Scientific
Phenolphthalein ≥98%
Katalog # A17135.30
CAS nummer:  77-09-8
Phenolphthalein ≥98%
Phenolphthalein ≥98%
Glass bottle for solids, 250 g
CAS nummer:  77-09-8
Price
675,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Purity: 98% CAS No.: 77-09-8 Molecular Formula: C20H14O4 Formula Weight: 318.33 Melting Point: 258-262° Beilstein Reference: 284423 Merck Reference: 14,7243 MDL No.: MFCD00005913
Synonymer:  3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
  • CAS nummer:77-09-8
  • Formel:C₂₀H₁₄O₄
  • MDL nummer:MFCD00005913
  • Molecular Weight:318.33 g/mol
  • Merck Index:14,7243
  • Smeltende Pt:258…262 °C
  • Tæthed:1,299
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-004-7
  • NÅ:01-2119498295-24
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g