DL-α-Tocopherol ≥97%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97+% CAS No.: 10191-41-0 Molecular Formula: C29H50O2 Formula Weight: 430.72 Flash Point: 240°(464°F) Physical Form: Viscous Liquid Density: 0.950 Refractive Index: 1.5050 Sensitivity: Light Sensitive Melting Point: 2-4° Boiling Point: 200-220°/0.1mm Beilstein Reference: 94012 Merck Reference: 14,10021 MDL No.: MFCD00072051
Synonyms: (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E
- CAS Number:10191-41-0
- Formula:C₂₉H₅₀O₂
- MDL Number:MFCD00072051
- Molecular Weight:430.71 g/mol
- Merck Index:14,10021
- Boiling Pt:200…220 °C (0,1 mmHg)
- Melting Pt:2…4 °C
- Density:0,950
- Flash Pt:240 °C (464 °F)
- Storage Temperature:Stue temperatur
- EINECS:233-466-0
Documents
Certificates
Search by Lot NumberALFAA17039.18_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:250 g



