Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tannin

Tannin

Glass bottle for solids, 250 g
Katalog # A17022.30
Leverandør:   Thermo Fisher Scientific
Tannin
Katalog # A17022.30
CAS nummer:  1401-55-4
Tannin
Tannin

Glass bottle for solids, 250 g

CAS nummer:  1401-55-4
Price
527,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

CAS No.: 1401-55-4 Molecular Formula: C76H52O46 Formula Weight: 1701.23 Odour: Odourless pH: 3.5 Melting Point: ca 218° dec. Beilstein Reference: 8186396 Merck Reference: 14,9052 MDL No.: MFCD00066397

Synonymer:  Gallotannin, Tannin
  • CAS nummer:1401-55-4
  • Formel:C₇₆H₅₂O₄₆
  • MDL nummer:MFCD00066397
  • Molecular Weight:1701.2 g/mol
  • Merck Index:14,9052
  • Smeltende Pt:∼ 218 °C (dec.)
  • Tæthed:>1 g/cm³ (20 °C)
  • Flash Pt:199 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:215-753-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.