Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tannin

Tannin

Glass bottle for solids, 1.000 g
Katalog # A17022.0B
Leverandør:   Thermo Fisher Scientific
Tannin
Katalog # A17022.0B
CAS nummer:  1401-55-4
Tannin
Tannin

Glass bottle for solids, 1.000 g

CAS nummer:  1401-55-4
Price
1.740,00 kr
Log ind for tilgængelighed
1 * 1.000 g
Product Details & Documents

CAS No.: 1401-55-4 Molecular Formula: C76H52O46 Formula Weight: 1701.23 Odour: Odourless pH: 3.5 Melting Point: ca 218° dec. Beilstein Reference: 8186396 Merck Reference: 14,9052 MDL No.: MFCD00066397

Synonymer:  Gallotannin, Tannin
  • CAS nummer:1401-55-4
  • Formel:C₇₆H₅₂O₄₆
  • MDL nummer:MFCD00066397
  • Molecular Weight:1701.2 g/mol
  • Merck Index:14,9052
  • Smeltende Pt:∼ 218 °C (dec.)
  • Tæthed:>1 g/cm³ (20 °C)
  • Flash Pt:199 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:215-753-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifikationer