Orders LinkContactUsLinkComponent
Xylitol ≥99%
Glass bottle for solids, 500 g
  A16944.36
Xylitol ≥99%
  A16944.36
 :  87-99-0
Xylitol ≥99%
Xylitol ≥99%
Glass bottle for solids, 500 g
 :  87-99-0
1.670,00 kr
1 * 500 g
Purity: 99% CAS No.: 87-99-0 Molecular Formula: C5H12O5 Formula Weight: 152.15 Odour: Odourless Melting Point: 92-96° Beilstein Reference: 1720523 Merck Reference: 14,10085 MDL No.: MFCD00064292
 :  (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
  •  :87-99-0
  •  :C₅H₁₂O₅
  •  :MFCD00064292
  •  :152.15 g/mol
  •  :14,10085
  •  :495 °C (1013 hPa)
  •  :92…96 °C
  •  :1,515
  •  :Stue temperatur
  •  :201-788-0
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g