Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Xylitol ≥99%

Xylitol ≥99%

Glass bottle for solids, 100 g
Katalog # A16944.22
Leverandør:   Thermo Fisher Scientific
Xylitol ≥99%
Katalog # A16944.22
CAS nummer:  87-99-0
Xylitol ≥99%
Xylitol ≥99%

Glass bottle for solids, 100 g

CAS nummer:  87-99-0
Price
500,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 87-99-0 Molecular Formula: C5H12O5 Formula Weight: 152.15 Odour: Odourless Melting Point: 92-96° Beilstein Reference: 1720523 Merck Reference: 14,10085 MDL No.: MFCD00064292

Synonymer:  (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
  • CAS nummer:87-99-0
  • Formel:C₅H₁₂O₅
  • MDL nummer:MFCD00064292
  • Molecular Weight:152.15 g/mol
  • Merck Index:14,10085
  • Kogende Pt:495 °C (1013 hPa)
  • Smeltende Pt:92…96 °C
  • Tæthed:1,515
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-788-0
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer