Bromphenol blå natriumsalt
Glass bottle for solids, 50 g
Leverandør: Thermo Fisher Scientific
Bromphenol blå natriumsalt
Katalog # A16899.18
Leverandør: Thermo Fisher Scientific
CAS nummer: 34725-61-6
Gå til:
Product Details & Documents
CAS No.: 34725-61-6 Molecular Formula: C19H9Br4NaO5S Formula Weight: 691.96 Melting Point: 300° Merck Reference: 14,1444 MDL No.: MFCD00013793
Synonymer: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt
- CAS nummer:34725-61-6
- Formel:C₁₉H₉Br₄NaO₅S
- MDL nummer:MFCD00013793
- Molecular Weight:691.95 g/mol
- Merck Index:14,1444
- Smeltende Pt:>300 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:252-170-2
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:50 g
- Cat. No.:A16899.18



