Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bromphenol blå natriumsalt

Bromphenol blå natriumsalt

Glass bottle for solids, 10 g
Katalog # A16899.09
Leverandør:   Thermo Fisher Scientific
Bromphenol blå natriumsalt
Katalog # A16899.09
CAS nummer:  34725-61-6
Bromphenol blå natriumsalt
Bromphenol blå natriumsalt

Glass bottle for solids, 10 g

CAS nummer:  34725-61-6
Price
427,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents

CAS No.: 34725-61-6 Molecular Formula: C19H9Br4NaO5S Formula Weight: 691.96 Melting Point: 300° Merck Reference: 14,1444 MDL No.: MFCD00013793

Synonymer:  Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt
  • CAS nummer:34725-61-6
  • Formel:C₁₉H₉Br₄NaO₅S
  • MDL nummer:MFCD00013793
  • Molecular Weight:691.95 g/mol
  • Merck Index:14,1444
  • Smeltende Pt:>300 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:252-170-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.