Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kaliumacetat ≥99%

Kaliumacetat ≥99%

Glass bottle for solids, 2.500 g
Katalog # A16321.0E
Leverandør:   Thermo Fisher Scientific
Kaliumacetat ≥99%
Katalog # A16321.0E
CAS nummer:  127-08-2
Kaliumacetat ≥99%
Kaliumacetat ≥99%

Glass bottle for solids, 2.500 g

CAS nummer:  127-08-2
Price
1.460,00 kr
Log ind for tilgængelighed
1 * 2.500 g
Product Details & Documents

Purity: 99% CAS No.: 127-08-2 Molecular Formula: CH3CO2K Formula Weight: 98.15 Melting Point: 292° Beilstein Reference: 3595449 Merck Reference: 14,7605 MDL No.: MFCD00012458

Synonymer:  Acetic acid potassium salt
  • CAS nummer:127-08-2
  • Formel:CH₃COOK
  • MDL nummer:MFCD00012458
  • Molecular Weight:98.14 g/mol
  • Merck Index:14,7605
  • Smeltende Pt:292 °C
  • Tæthed:1,57
  • Flash Pt:>250 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-822-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2.500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.