MES monohydrat ≥98%
Glass bottle for solids, 500 g
Purity: 98% CAS No.: 145224-94-8 Molecular Formula: C6H13NO4S.H2O Formula Weight: 213.26 (195.24anhy) Physical Form: Crystalline powder Appearance: White Melting Point: ca 308° dec. Beilstein Reference: 141319 Merck Reference: 14,5902 MDL No.: MFCD00149409
Good's buffer
: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
- :145224-94-8
- :C₆H₁₃NO₄S·1H₂O
- :MFCD00149409
- :213.25 g/mol
- :14,5902
- :∼ 308 °C (dec.)
- :Stue temperatur
- :224-632-3
- Pakning:Glass bottle for solids
- Pakn:500 g