Orders LinkContactUsLinkComponent
MES monohydrat ≥98%
Glass bottle for solids, 500 g
  A16104.36
MES monohydrat ≥98%
  A16104.36
 :  145224-94-8
MES monohydrat ≥98%
MES monohydrat ≥98%
Glass bottle for solids, 500 g
 :  145224-94-8
3.890,00 kr
1 * 500 g
Purity: 98% CAS No.: 145224-94-8 Molecular Formula: C6H13NO4S.H2O Formula Weight: 213.26 (195.24anhy) Physical Form: Crystalline powder Appearance: White Melting Point: ca 308° dec. Beilstein Reference: 141319 Merck Reference: 14,5902 MDL No.: MFCD00149409
Good's buffer
 :  MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
  •  :145224-94-8
  •  :C₆H₁₃NO₄S·1H₂O
  •  :MFCD00149409
  •  :213.25 g/mol
  •  :14,5902
  •  :∼ 308 °C (dec.)
  •  :Stue temperatur
  •  :224-632-3
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g