Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
MES monohydrat ≥98%

MES monohydrat ≥98%

Glass bottle for solids, 100 g
Katalog # A16104.22
Irriterende
Leverandør:   Thermo Fisher Scientific
MES monohydrat ≥98%
Katalog # A16104.22
CAS nummer:  145224-94-8
MES monohydrat ≥98%
MES monohydrat ≥98%

Glass bottle for solids, 100 g

CAS nummer:  145224-94-8
Price
1.080,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 145224-94-8 Molecular Formula: C6H13NO4S.H2O Formula Weight: 213.26 (195.24anhy) Physical Form: Crystalline powder Appearance: White Melting Point: ca 308° dec. Beilstein Reference: 141319 Merck Reference: 14,5902 MDL No.: MFCD00149409

Good's buffer

Synonymer:  MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
  • CAS nummer:145224-94-8
  • Formel:C₆H₁₃NO₄S·1H₂O
  • MDL nummer:MFCD00149409
  • Molecular Weight:213.25 g/mol
  • Merck Index:14,5902
  • Smeltende Pt:∼ 308 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:224-632-3
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.