Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

MES monohydrat ≥98%

Glass bottle for solids, 25 g
Katalog # A16104.14
Irriterende
Leverandør:   Thermo Fisher Scientific
MES monohydrat ≥98%
Katalog # A16104.14
CAS nummer:  145224-94-8
MES monohydrat ≥98%
MES monohydrat ≥98%

Glass bottle for solids, 25 g

CAS nummer:  145224-94-8
Price
378,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 145224-94-8 Molecular Formula: C6H13NO4S.H2O Formula Weight: 213.26 (195.24anhy) Physical Form: Crystalline powder Appearance: White Melting Point: ca 308° dec. Beilstein Reference: 141319 Merck Reference: 14,5902 MDL No.: MFCD00149409

Good's buffer

Synonymer:  MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
  • CAS nummer:145224-94-8
  • Formel:C₆H₁₃NO₄S·1H₂O
  • MDL nummer:MFCD00149409
  • Molecular Weight:213.25 g/mol
  • Merck Index:14,5902
  • Smeltende Pt:∼ 308 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:224-632-3
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer