Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Thiomorpholin ≥98%

Thiomorpholin ≥98%

Glass bottle for solids, 100 g
Katalog # A15958.22
Ætsende
Leverandør:   Thermo Fisher Scientific
Thiomorpholin ≥98%
Katalog # A15958.22
CAS nummer:  123-90-0
Thiomorpholin ≥98%
Thiomorpholin ≥98%

Glass bottle for solids, 100 g

CAS nummer:  123-90-0
Price
9.030,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 123-90-0 Molecular Formula: C4H9NS Formula Weight: 103.19 Flash Point: 67°(153°F) Refractive Index: 1.5384 Boiling Point: 175-177° Solubility: Miscible with water Beilstein Reference: 102550 Merck Reference: 14,9300 UN No.: 3267 MDL No.: MFCD00005974

  • CAS nummer:123-90-0
  • Formel:C₄H₉NS
  • MDL nummer:MFCD00005974
  • Molecular Weight:103.19 g/mol
  • Merck Index:14,9300
  • Kogende Pt:175…177 °C
  • Tæthed:1,086
  • Flash Pt:67 °C (153 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-660-2
  • FN:3267
  • ADR:8,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.