Orders LinkContactUsLinkComponent
Thiomorpholin ≥98%
Thiomorpholin ≥98%
Glass bottle for solids, 25 g
  A15958.14
Thiomorpholin ≥98%
  A15958.14
 :  123-90-0
Thiomorpholin ≥98%
Thiomorpholin ≥98%
Glass bottle for solids, 25 g
 :  123-90-0
2.480,00 kr
1 * 25 g
Purity: 98% CAS No.: 123-90-0 Molecular Formula: C4H9NS Formula Weight: 103.19 Flash Point: 67°(153°F) Refractive Index: 1.5384 Boiling Point: 175-177° Solubility: Miscible with water Beilstein Reference: 102550 Merck Reference: 14,9300 UN No.: 3267 MDL No.: MFCD00005974
  •  :123-90-0
  •  :C₄H₉NS
  •  :MFCD00005974
  •  :103.19 g/mol
  •  :14,9300
  •  :175…177 °C
  •  :1,086
  •  :67 °C (153 °F)
  •  :Stue temperatur
  •  :204-660-2
  •  :3267
  •  :8,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g