Thiomorpholin ≥98%
Glass bottle for solids, 25 g
Purity: 98% CAS No.: 123-90-0 Molecular Formula: C4H9NS Formula Weight: 103.19 Flash Point: 67°(153°F) Refractive Index: 1.5384 Boiling Point: 175-177° Solubility: Miscible with water Beilstein Reference: 102550 Merck Reference: 14,9300 UN No.: 3267 MDL No.: MFCD00005974
- :123-90-0
- :C₄H₉NS
- :MFCD00005974
- :103.19 g/mol
- :14,9300
- :175…177 °C
- :1,086
- :67 °C (153 °F)
- :Stue temperatur
- :204-660-2
- :3267
- :8,III
Documents
- Pakning:Glass bottle for solids
- Pakn:25 g