Thiomorpholin ≥98%
Glass bottle for solids, 5 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 98% CAS No.: 123-90-0 Molecular Formula: C4H9NS Formula Weight: 103.19 Flash Point: 67°(153°F) Refractive Index: 1.5384 Boiling Point: 175-177° Solubility: Miscible with water Beilstein Reference: 102550 Merck Reference: 14,9300 UN No.: 3267 MDL No.: MFCD00005974
- CAS nummer:123-90-0
- Formel:C₄H₉NS
- MDL nummer:MFCD00005974
- Molecular Weight:103.19 g/mol
- Merck Index:14,9300
- Kogende Pt:175…177 °C
- Tæthed:1,086
- Flash Pt:67 °C (153 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-660-2
- FN:3267
- ADR:8,III
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



