Orders LinkContactUsLinkComponent
2-Mercaptoethanol ≥98%
Glass bottle for solids, 5.000 g
Katalog # A15890.0I
Giftig
Ætsende
Irriterende
Leverandør:   Thermo Fisher Scientific
2-Mercaptoethanol ≥98%
Katalog # A15890.0I
CAS nummer:  60-24-2
2-Mercaptoethanol ≥98%
2-Mercaptoethanol ≥98%
Glass bottle for solids, 5.000 g
CAS nummer:  60-24-2
Price
3.800,00 kr
Log ind for tilgængelighed
1 * 5.000 g
Product Details & Documents
Purity: 98+% CAS No.: 60-24-2 Molecular Formula: HSCH2CH2OH Formula Weight: 78.13 Flash Point: 73°(163°F) Refractive Index: 1.500 Odour: Unpleasant Melting Point: -100° Boiling Point: 156-158° Beilstein Reference: 773648 Merck Reference: 14,5869 UN No.: 2966 MDL No.: MFCD00004890
Synonymer:  2-Hydroxyethyl mercaptan, β-Mercaptoethanol
  • CAS nummer:60-24-2
  • Formel:HSCH₂CH₂OH
  • MDL nummer:MFCD00004890
  • Molecular Weight:78.14 g/mol
  • Merck Index:14,5869
  • Kogende Pt:156…158 °C
  • Smeltende Pt:–100 °C
  • Tæthed:1,114
  • Flash Pt:73 °C (163 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-464-6
  • FN:2966
  • ADR:6.1,II
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    5.000 g
Specifikationer