Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(2-Hydroxyethyl)trimethylammoniumchlorid ≥98%

(2-Hydroxyethyl)trimethylammoniumchlorid ≥98%

Glass bottle for solids, 500 g
Katalog # A15828.36
Irriterende
Leverandør:   Thermo Fisher Scientific
(2-Hydroxyethyl)trimethylammoniumchlorid ≥98%
Katalog # A15828.36
CAS nummer:  67-48-1
(2-Hydroxyethyl)trimethylammoniumchlorid ≥98%
(2-Hydroxyethyl)trimethylammoniumchlorid ≥98%

Glass bottle for solids, 500 g

CAS nummer:  67-48-1
Price
784,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 67-48-1 Molecular Formula: HOCH2CH2N(CH3)3Cl Formula Weight: 139.63 Physical Form: Solid Sensitivity: Hygroscopic Beilstein Reference: 3563126 Merck Reference: 14,2206 MDL No.: MFCD00011721

An acyl group acceptor

Synonymer:  2-hydroxy-N,N,N-trimethylethanaminium chloride
  • CAS nummer:67-48-1
  • Formel:HOH₂CCH₂N(CH₃)₃Cl
  • MDL nummer:MFCD00011721
  • Molecular Weight:139.62 g/mol
  • Merck Index:14,2206
  • Smeltende Pt:∼ 305 °C (dec.)
  • Tæthed:∼1,205 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-655-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.