Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

(2-Hydroxyethyl)trimethylammoniumchlorid ≥98%

Glass bottle for solids, 2.500 g
Katalog # A15828.0E
Irriterende
Leverandør:   Thermo Fisher Scientific
(2-Hydroxyethyl)trimethylammoniumchlorid ≥98%
Katalog # A15828.0E
CAS nummer:  67-48-1
(2-Hydroxyethyl)trimethylammoniumchlorid ≥98%
(2-Hydroxyethyl)trimethylammoniumchlorid ≥98%

Glass bottle for solids, 2.500 g

CAS nummer:  67-48-1
Price
2.960,00 kr
Log ind for tilgængelighed
1 * 2.500 g
Product Details & Documents

Purity: 98+% CAS No.: 67-48-1 Molecular Formula: HOCH2CH2N(CH3)3Cl Formula Weight: 139.63 Physical Form: Solid Sensitivity: Hygroscopic Beilstein Reference: 3563126 Merck Reference: 14,2206 MDL No.: MFCD00011721

An acyl group acceptor

Synonymer:  2-hydroxy-N,N,N-trimethylethanaminium chloride
  • CAS nummer:67-48-1
  • Formel:HOH₂CCH₂N(CH₃)₃Cl
  • MDL nummer:MFCD00011721
  • Molecular Weight:139.62 g/mol
  • Merck Index:14,2206
  • Smeltende Pt:∼ 305 °C (dec.)
  • Tæthed:∼1,205 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-655-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2.500 g

Specifikationer