Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L-Alanin ≥99%

L-Alanin ≥99%

Glass bottle for solids, 500 g
Katalog # A15804.36
Leverandør:   Thermo Fisher Scientific
L-Alanin ≥99%
Katalog # A15804.36
CAS nummer:  56-41-7
L-Alanin ≥99%
L-Alanin ≥99%

Glass bottle for solids, 500 g

CAS nummer:  56-41-7
Price
3.710,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 56-41-7 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Melting Point: ca 295° dec. Beilstein Reference: 1720248 Merck Reference: 14,204 Optical Rotation: +14° (c=5 in 5N HCl) MDL No.: MFCD00064410

Synonymer:  (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
  • CAS nummer:56-41-7
  • Formel:C₃H₇NO₂
  • MDL nummer:MFCD00064410
  • Molecular Weight:89.09 g/mol
  • Merck Index:14,204
  • Kogende Pt:189 °C (1013 hPa)
  • Smeltende Pt:∼ 295 °C (dec.)
  • Tæthed:1,432
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-273-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.