Orders LinkContactUsLinkComponent
L-Alanin ≥99%
L-Alanin ≥99%
Glass bottle for solids, 25 g
  A15804.14
L-Alanin ≥99%
  A15804.14
 :  56-41-7
L-Alanin ≥99%
L-Alanin ≥99%
Glass bottle for solids, 25 g
 :  56-41-7
403,00 kr
1 * 25 g
Purity: 99% CAS No.: 56-41-7 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Melting Point: ca 295° dec. Beilstein Reference: 1720248 Merck Reference: 14,204 Optical Rotation: +14° (c=5 in 5N HCl) MDL No.: MFCD00064410
 :  (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
  •  :56-41-7
  •  :C₃H₇NO₂
  •  :MFCD00064410
  •  :89.09 g/mol
  •  :14,204
  •  :189 °C (1013 hPa)
  •  :∼ 295 °C (dec.)
  •  :1,432
  •  :Stue temperatur
  •  :200-273-8
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g