L-Alanin ≥99%
Glass bottle for solids, 25 g
Purity: 99% CAS No.: 56-41-7 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Melting Point: ca 295° dec. Beilstein Reference: 1720248 Merck Reference: 14,204 Optical Rotation: +14° (c=5 in 5N HCl) MDL No.: MFCD00064410
: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
- :56-41-7
- :C₃H₇NO₂
- :MFCD00064410
- :89.09 g/mol
- :14,204
- :189 °C (1013 hPa)
- :∼ 295 °C (dec.)
- :1,432
- :Stue temperatur
- :200-273-8
Documents
- Pakning:Glass bottle for solids
- Pakn:25 g