Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
D(+)-Sucrose ≥99%

D(+)-Sucrose ≥99%

Glass bottle for solids, 10.000 g
Katalog # A15583.0C
Leverandør:   Thermo Fisher Scientific
D(+)-Sucrose ≥99%
Katalog # A15583.0C
CAS nummer:  57-50-1
D(+)-Sucrose ≥99%
D(+)-Sucrose ≥99%

Glass bottle for solids, 10.000 g

CAS nummer:  57-50-1
Price
2.710,00 kr
Log ind for tilgængelighed
1 * 10.000 g
Product Details & Documents

Purity: 99% CAS No.: 57-50-1 Molecular Formula: C12H22O11 Formula Weight: 342.30 Odour: Odourless Melting Point: 186-190° Beilstein Reference: 90825 Merck Reference: 14,8881 Optical Rotation: +66.5° (c=10 in water) MDL No.: MFCD00006626

Synonymer:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS nummer:57-50-1
  • Formel:C₁₂H₂₂O₁₁
  • MDL nummer:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck Index:14,8881
  • Smeltende Pt:186…190 °C
  • Tæthed:1,580
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-334-9
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10.000 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.