Orders LinkContactUsLinkComponent
Hydroxylammoniumchlorid 99%
Hydroxylammoniumchlorid 99%
Glass bottle for solids, 100 g
Katalog # A15398.22
Sundhed
Ætsende
Irriterende
Miljø
Leverandør:   Thermo Fisher Scientific
Hydroxylammoniumchlorid 99%
Katalog # A15398.22
CAS nummer:  5470-11-1
Hydroxylammoniumchlorid 99%
Hydroxylammoniumchlorid 99%
Glass bottle for solids, 100 g
CAS nummer:  5470-11-1
Price
385,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Purity: 99% CAS No.: 5470-11-1 Molecular Formula: H3NO.HCl Formula Weight: 69.49 Odour: Acidic pH: 3.6 Melting Point: 152° dec. Beilstein Reference: 3539763 Merck Reference: 14,4828 UN No.: 2923 MDL No.: MFCD00051089
Synonymer:  Hydroxylamine HCl, Hydroxylamine hydrochloride
  • CAS nummer:5470-11-1
  • Formel:NH₂OH·HCl
  • MDL nummer:MFCD00051089
  • Molecular Weight:69.49 g/mol
  • Merck Index:14,4828
  • Kogende Pt:306 °C (1013 hPa)
  • Smeltende Pt:152 °C (dec.)
  • Tæthed:1,67
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:226-798-2
  • FN:2923
  • ADR:8,III
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g