Orders LinkContactUsLinkComponent
Hydroxylammoniumchlorid 99%
Hydroxylammoniumchlorid 99%
Glass bottle for solids, 5.000 g
Catalog # A15398.0I
Health
Corrosive
Irritant
Environment
Hydroxylammoniumchlorid 99%
Catalog # A15398.0I
CAS Number:  5470-11-1
Hydroxylammoniumchlorid 99%
Hydroxylammoniumchlorid 99%
Glass bottle for solids, 5.000 g
CAS Number:  5470-11-1
Price
6.550,00 kr
Log in for availability
1 * 5.000 g
Product Details & Documents
Purity: 99% CAS No.: 5470-11-1 Molecular Formula: H3NO.HCl Formula Weight: 69.49 Odour: Acidic pH: 3.6 Melting Point: 152° dec. Beilstein Reference: 3539763 Merck Reference: 14,4828 UN No.: 2923 MDL No.: MFCD00051089
Synonyms:  Hydroxylamine HCl, Hydroxylamine hydrochloride
  • CAS Number:5470-11-1
  • Formula:NH₂OH·HCl
  • MDL Number:MFCD00051089
  • Molecular Weight:69.49 g/mol
  • Merck Index:14,4828
  • Boiling Pt:306 °C (1013 hPa)
  • Melting Pt:152 °C (dec.)
  • Density:1,67
  • Storage Temperature:Stue temperatur
  • EINECS:226-798-2
  • UN:2923
  • ADR:8,III
Specifications
Specifications
  • Pakning:
    Glass bottle for solids
  • Pakn:
    5.000 g