Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ammoniumchlorid ≥98%

Ammoniumchlorid ≥98%

Glass bottle for solids, 500 g
Katalog # A15000.36
Irriterende
Leverandør:   Thermo Fisher Scientific
Ammoniumchlorid ≥98%
Katalog # A15000.36
CAS nummer:  12125-02-9
Ammoniumchlorid ≥98%
Ammoniumchlorid ≥98%

Glass bottle for solids, 500 g

CAS nummer:  12125-02-9
Price
427,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 12125-02-9 Molecular Formula: H4ClN Formula Weight: 53.49 Refractive Index: 1.642 Odour: Odourless Beilstein Reference: 4371014 Merck Reference: 14,509 MDL No.: MFCD00011420

Synonymer:  Ammonium muriate, Salmiac
  • CAS nummer:12125-02-9
  • Formel:NH₄Cl
  • MDL nummer:MFCD00011420
  • Molecular Weight:53.49 g/mol
  • Merck Index:14,509
  • Smeltende Pt:340 °C (subl.)
  • Tæthed:1,53
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:235-186-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer