Ammoniumchlorid ≥98%
Glass bottle for solids, 250 g
Purity: 98+% CAS No.: 12125-02-9 Molecular Formula: H4ClN Formula Weight: 53.49 Refractive Index: 1.642 Odour: Odourless Beilstein Reference: 4371014 Merck Reference: 14,509 MDL No.: MFCD00011420
: Ammonium muriate, Salmiac
- :12125-02-9
- :NH₄Cl
- :MFCD00011420
- :53.49 g/mol
- :14,509
- :340 °C (subl.)
- :1,53
- :Stue temperatur
- :235-186-4
Documents
- Pakning:Glass bottle for solids
- Pakn:250 g
- Cat. No.:A15000.30
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



