Orders LinkContactUsLinkComponent
Ammoniumchlorid ≥98%
Glass bottle for solids, 1.000 g
  A15000.0B
Ammoniumchlorid ≥98%
  A15000.0B
 :  12125-02-9
Ammoniumchlorid ≥98%
Ammoniumchlorid ≥98%
Glass bottle for solids, 1.000 g
 :  12125-02-9
654,00 kr
1 * 1.000 g
Purity: 98+% CAS No.: 12125-02-9 Molecular Formula: H4ClN Formula Weight: 53.49 Refractive Index: 1.642 Odour: Odourless Beilstein Reference: 4371014 Merck Reference: 14,509 MDL No.: MFCD00011420
 :  Ammonium muriate, Salmiac
  •  :12125-02-9
  •  :NH₄Cl
  •  :MFCD00011420
  •  :53.49 g/mol
  •  :14,509
  •  :340 °C (subl.)
  •  :1,53
  •  :Stue temperatur
  •  :235-186-4
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g