Ammoniumchlorid ≥98%
Glass bottle for solids, 1.000 g
Purity: 98+% CAS No.: 12125-02-9 Molecular Formula: H4ClN Formula Weight: 53.49 Refractive Index: 1.642 Odour: Odourless Beilstein Reference: 4371014 Merck Reference: 14,509 MDL No.: MFCD00011420
: Ammonium muriate, Salmiac
- :12125-02-9
- :NH₄Cl
- :MFCD00011420
- :53.49 g/mol
- :14,509
- :340 °C (subl.)
- :1,53
- :Stue temperatur
- :235-186-4
- Pakning:Glass bottle for solids
- Pakn:1.000 g