2-(4-Hydroxyphenyl)ethanol ≥98%
Glass bottle for solids, 5 g
Leverandør: Thermo Fisher Scientific
2-(4-Hydroxyphenyl)ethanol ≥98%
Katalog # A14891.06
Leverandør: Thermo Fisher Scientific
CAS nummer: 501-94-0
Gå til:
Product Details & Documents
Purity: 98% CAS No.: 501-94-0 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 89-92° Boiling Point: 139°/1mm MDL No.: MFCD00002902
Synonymer: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol
- CAS nummer:501-94-0
- Formel:C₈H₁₀O₂
- MDL nummer:MFCD00002902
- Molecular Weight:138.17 g/mol
- Kogende Pt:139 °C (1 mmHg)
- Smeltende Pt:89…92 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:207-930-8
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g



