2-(4-Hydroxyphenyl)ethanol ≥98%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
2-(4-Hydroxyphenyl)ethanol ≥98%
Catalog # A14891.03
Supplier: Thermo Fisher Scientific
CAS Number: 501-94-0
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 501-94-0 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 89-92° Boiling Point: 139°/1mm MDL No.: MFCD00002902
Synonyms: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol
- CAS Number:501-94-0
- Formula:C₈H₁₀O₂
- MDL Number:MFCD00002902
- Molecular Weight:138.17 g/mol
- Boiling Pt:139 °C (1 mmHg)
- Melting Pt:89…92 °C
- Storage Temperature:Stue temperatur
- EINECS:207-930-8
Documents
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:1 g
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



