2-Hexanon ≥98%
Glass bottle for solids, 500 g
Purity: 98% CAS No.: 591-78-6 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 23°(73°F) Refractive Index: 1.4010 Odour: Pungent Melting Point: -56° Boiling Point: 126-127° Beilstein Reference: 1737676 Merck Reference: 14,6033 UN No.: 1224 MDL No.: MFCD00009482
: Butylmethylketon
- :591-78-6
- :C₆H₁₂O
- :MFCD00009482
- :100.16 g/mol
- :14,6033
- :126…127 °C
- :–56 °C
- :0,811
- :23 °C (73 °F)
- :Stue temperatur
- :209-731-1
- :1224
- :3,III
- Pakning:Glass bottle for solids
- Pakn:500 g