2-Hexanon ≥98%
Glass bottle for solids, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 98% CAS No.: 591-78-6 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 23°(73°F) Refractive Index: 1.4010 Odour: Pungent Melting Point: -56° Boiling Point: 126-127° Beilstein Reference: 1737676 Merck Reference: 14,6033 UN No.: 1224 MDL No.: MFCD00009482
Synonymer: Butylmethylketon
- CAS nummer:591-78-6
- Formel:C₆H₁₂O
- MDL nummer:MFCD00009482
- Molecular Weight:100.16 g/mol
- Merck Index:14,6033
- Kogende Pt:126…127 °C
- Smeltende Pt:–56 °C
- Tæthed:0,811
- Flash Pt:23 °C (73 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:209-731-1
- FN:1224
- ADR:3,III
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g



