Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Hexanon ≥98%

2-Hexanon ≥98%

Glass bottle for solids, 50 g
Catalog # A14867.18
Health
Flammable
Irritant
2-Hexanon ≥98%
Catalog # A14867.18
CAS Number:  591-78-6
2-Hexanon ≥98%
2-Hexanon ≥98%

Glass bottle for solids, 50 g

CAS Number:  591-78-6
Price
903,00 kr
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 98% CAS No.: 591-78-6 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 23°(73°F) Refractive Index: 1.4010 Odour: Pungent Melting Point: -56° Boiling Point: 126-127° Beilstein Reference: 1737676 Merck Reference: 14,6033 UN No.: 1224 MDL No.: MFCD00009482

Synonyms:  Butylmethylketon
  • CAS Number:591-78-6
  • Formula:C₆H₁₂O
  • MDL Number:MFCD00009482
  • Molecular Weight:100.16 g/mol
  • Merck Index:14,6033
  • Boiling Pt:126…127 °C
  • Melting Pt:–56 °C
  • Density:0,811
  • Flash Pt:23 °C (73 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:209-731-1
  • UN:1224
  • ADR:3,III
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.