Orders LinkContactUsLinkComponent
2-Hexanon ≥98%
Glass bottle for solids, 10 g
  A14867.09
2-Hexanon ≥98%
  A14867.09
 :  591-78-6
2-Hexanon ≥98%
2-Hexanon ≥98%
Glass bottle for solids, 10 g
 :  591-78-6
366,00 kr
1 * 10 g
Purity: 98% CAS No.: 591-78-6 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 23°(73°F) Refractive Index: 1.4010 Odour: Pungent Melting Point: -56° Boiling Point: 126-127° Beilstein Reference: 1737676 Merck Reference: 14,6033 UN No.: 1224 MDL No.: MFCD00009482
 :  Butylmethylketon
  •  :591-78-6
  •  :C₆H₁₂O
  •  :MFCD00009482
  •  :100.16 g/mol
  •  :14,6033
  •  :126…127 °C
  •  :–56 °C
  •  :0,811
  •  :23 °C (73 °F)
  •  :Stue temperatur
  •  :209-731-1
  •  :1224
  •  :3,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g