Caprinsyre ≥99%
Glass bottle for solids, 250 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 334-48-5 Molecular Formula: CH3(CH2)8CO2H Formula Weight: 172.27 Flash Point: 147°(296°F) Physical Form: Cryst solid Density: 0.893 Appearance: White pH: 4 Melting Point: 29-33° Boiling Point: 268-270° Beilstein Reference: 1754556 Merck Reference: 14,1758 MDL No.: MFCD00004441
Synonymer: Decylic acid, Decanoic acid
- CAS nummer:334-48-5
- Formel:C₁₀H₂₀O₂
- MDL nummer:MFCD00004441
- Molecular Weight:172.27 g/mol
- Merck Index:14,1758
- Kogende Pt:268…270 °C
- Smeltende Pt:29…33 °C
- Tæthed:0,893
- Flash Pt:147 °C (296 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:206-376-4
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 g



