Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Caprinsyre ≥99%

Caprinsyre ≥99%

Glass bottle for solids, 5.000 g
Katalog # A14788.0I
Irriterende
Leverandør:   Thermo Fisher Scientific
Caprinsyre ≥99%
Katalog # A14788.0I
CAS nummer:  334-48-5
Caprinsyre ≥99%
Caprinsyre ≥99%

Glass bottle for solids, 5.000 g

CAS nummer:  334-48-5
Price
4.620,00 kr
Log ind for tilgængelighed
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 334-48-5 Molecular Formula: CH3(CH2)8CO2H Formula Weight: 172.27 Flash Point: 147°(296°F) Physical Form: Cryst solid Density: 0.893 Appearance: White pH: 4 Melting Point: 29-33° Boiling Point: 268-270° Beilstein Reference: 1754556 Merck Reference: 14,1758 MDL No.: MFCD00004441

Synonymer:  Decylic acid, Decanoic acid
  • CAS nummer:334-48-5
  • Formel:C₁₀H₂₀O₂
  • MDL nummer:MFCD00004441
  • Molecular Weight:172.27 g/mol
  • Merck Index:14,1758
  • Kogende Pt:268…270 °C
  • Smeltende Pt:29…33 °C
  • Tæthed:0,893
  • Flash Pt:147 °C (296 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:206-376-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5.000 g

Specifikationer