Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Diethylenglycol ≥99%

Diethylenglycol ≥99%

Glass bottle for solids, 250 g
Katalog # A14728.30
Irriterende
Leverandør:   Thermo Fisher Scientific
Diethylenglycol ≥99%
Katalog # A14728.30
CAS nummer:  111-46-6
Diethylenglycol ≥99%
Diethylenglycol ≥99%

Glass bottle for solids, 250 g

CAS nummer:  111-46-6
Price
357,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 111-46-6 Molecular Formula: C4H10O3 Formula Weight: 106.12 Flash Point: 143°(289°F) Physical Form: Clear liquid Density: 1.118 Refractive Index: 1.4470 Sensitivity: Hygroscopic Appearance: Colourless Odour: Nearly Odourless Melting Point: -10° Boiling Point: 245-246° Beilstein Reference: 969209 Merck Reference: 14,3119 MDL No.: MFCD00002882

Synonymer:  2,2'-Oxydiethanol
  • CAS nummer:111-46-6
  • Formel:C₄H₁₀O₃
  • MDL nummer:MFCD00002882
  • Molecular Weight:106.12 g/mol
  • Merck Index:14,3119
  • Kogende Pt:245…246 °C
  • Smeltende Pt:–10 °C
  • Tæthed:1,118
  • Flash Pt:143 °C (289 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-872-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.