Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Diethylenglycol ≥99%

Diethylenglycol ≥99%

Glass bottle for solids, 2.500 g
Katalog # A14728.0E
Irriterende
Leverandør:   Thermo Fisher Scientific
Diethylenglycol ≥99%
Katalog # A14728.0E
CAS nummer:  111-46-6
Diethylenglycol ≥99%
Diethylenglycol ≥99%

Glass bottle for solids, 2.500 g

CAS nummer:  111-46-6
Price
838,00 kr
Log ind for tilgængelighed
1 * 2.500 g
Product Details & Documents

Purity: 99% CAS No.: 111-46-6 Molecular Formula: C4H10O3 Formula Weight: 106.12 Flash Point: 143°(289°F) Physical Form: Clear liquid Density: 1.118 Refractive Index: 1.4470 Sensitivity: Hygroscopic Appearance: Colourless Odour: Nearly Odourless Melting Point: -10° Boiling Point: 245-246° Beilstein Reference: 969209 Merck Reference: 14,3119 MDL No.: MFCD00002882

Synonymer:  2,2'-Oxydiethanol
  • CAS nummer:111-46-6
  • Formel:C₄H₁₀O₃
  • MDL nummer:MFCD00002882
  • Molecular Weight:106.12 g/mol
  • Merck Index:14,3119
  • Kogende Pt:245…246 °C
  • Smeltende Pt:–10 °C
  • Tæthed:1,118
  • Flash Pt:143 °C (289 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-872-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2.500 g

Specifikationer