Orders LinkContactUsLinkComponent
Diethylenglycol ≥99%
Glass bottle for solids, 10.000 g
  A14728.0C
Diethylenglycol ≥99%
  A14728.0C
 :  111-46-6
Diethylenglycol ≥99%
Diethylenglycol ≥99%
Glass bottle for solids, 10.000 g
 :  111-46-6
2.760,00 kr
1 * 10.000 g
Purity: 99% CAS No.: 111-46-6 Molecular Formula: C4H10O3 Formula Weight: 106.12 Flash Point: 143°(289°F) Physical Form: Clear liquid Density: 1.118 Refractive Index: 1.4470 Sensitivity: Hygroscopic Appearance: Colourless Odour: Nearly Odourless Melting Point: -10° Boiling Point: 245-246° Beilstein Reference: 969209 Merck Reference: 14,3119 MDL No.: MFCD00002882
 :  2,2'-Oxydiethanol
  •  :111-46-6
  •  :C₄H₁₀O₃
  •  :MFCD00002882
  •  :106.12 g/mol
  •  :14,3119
  •  :245…246 °C
  •  :–10 °C
  •  :1,118
  •  :143 °C (289 °F)
  •  :Stue temperatur
  •  :203-872-2
  • Pakning:
    Glass bottle for solids
  • Pakn:
    10.000 g