Diethylenglycol ≥99%
Glass bottle for solids, 10.000 g
Purity: 99% CAS No.: 111-46-6 Molecular Formula: C4H10O3 Formula Weight: 106.12 Flash Point: 143°(289°F) Physical Form: Clear liquid Density: 1.118 Refractive Index: 1.4470 Sensitivity: Hygroscopic Appearance: Colourless Odour: Nearly Odourless Melting Point: -10° Boiling Point: 245-246° Beilstein Reference: 969209 Merck Reference: 14,3119 MDL No.: MFCD00002882
: 2,2'-Oxydiethanol
- :111-46-6
- :C₄H₁₀O₃
- :MFCD00002882
- :106.12 g/mol
- :14,3119
- :245…246 °C
- :–10 °C
- :1,118
- :143 °C (289 °F)
- :Stue temperatur
- :203-872-2
- Pakning:Glass bottle for solids
- Pakn:10.000 g