Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Iodacetamid 98% stabilised

Glass bottle for solids, 100 g
Katalog # A14715.22
Giftig
Irriterende
Leverandør:   Thermo Fisher Scientific
Iodacetamid 98% stabilised
Katalog # A14715.22
CAS nummer:  144-48-9
Iodacetamid 98% stabilised
Iodacetamid 98% stabilised

Glass bottle for solids, 100 g

CAS nummer:  144-48-9
Price
3.760,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation.

2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.

Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation

Synonymer:  2-Iodoacetamide
  • CAS nummer:144-48-9
  • Formel:ICH₂CONH₂
  • MDL nummer:MFCD00008028
  • Molecular Weight:184.96 g/mol
  • Kogende Pt:266 °C (1013 hPa)
  • Smeltende Pt:92…95 °C (dried)
  • Opbevaringstemperatur:Køleskab
  • EINECS:205-630-1
  • FN:2811
  • ADR:6.1,III
  •  :fugtet med vand ∼5-8%
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.