Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Iodacetamid 98% stabilised

Glass bottle for solids, 5 g
Catalog # A14715.06
Toxic
Irritant
Iodacetamid 98% stabilised
Catalog # A14715.06
CAS Number:  144-48-9
Iodacetamid 98% stabilised
Iodacetamid 98% stabilised

Glass bottle for solids, 5 g

CAS Number:  144-48-9
Price
kr337.00
Log in for availability
1 * 5 g
Product Details & Documents

Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation.

2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.

Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation

Synonyms:  2-Iodoacetamide
  • CAS Number:144-48-9
  • Formula:ICH₂CONH₂
  • MDL Number:MFCD00008028
  • Molecular Weight:184.96 g/mol
  • Boiling Pt:266 °C (1013 hPa)
  • Melting Pt:92…95 °C (dried)
  • Storage Temperature:Køleskab
  • EINECS:205-630-1
  • UN:2811
  • ADR:6.1,III
  •  :fugtet med vand ∼5-8%
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifications