Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tricine ≥98%

Tricine ≥98%

Glass bottle for solids, 25 g
Katalog # A14695.14
Leverandør:   Thermo Fisher Scientific
Tricine ≥98%
Katalog # A14695.14
CAS nummer:  5704-04-1
Tricine ≥98%
Tricine ≥98%

Glass bottle for solids, 25 g

CAS nummer:  5704-04-1
Price
349,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 5704-04-1 Molecular Formula: (HOCH2)3CNHCH2CO2H Formula Weight: 179.17 Melting Point: ca 183° dec. Beilstein Reference: 1937804 Merck Reference: 14,9651 MDL No.: MFCD00004277

Synonymer:  2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
  • CAS nummer:5704-04-1
  • Formel:(HOCH₂)₃CNHCH₂CO₂H
  • MDL nummer:MFCD00004277
  • Molecular Weight:179.17 g/mol
  • Merck Index:14,9651
  • Smeltende Pt:∼ 183 °C (dec.)
  • Tæthed:1,523 g/cm³ (19.85 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:227-193-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer