D-(+)-Cellobiose ≥98%
Glass bottle for solids, 5 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 98+% CAS No.: 528-50-7 Molecular Formula: C12H22O11 Formula Weight: 342.30 Physical Form: Powder Appearance: White Melting Point: 234° dec. Beilstein Reference: 93795 Merck Reference: 14,1961 Optical Rotation: +34° (c=10 in water, 15h) MDL No.: MFCD00136034
Synonymer: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
- CAS nummer:528-50-7
- Formel:C₁₂H₂₂O₁₁
- MDL nummer:MFCD00136034
- Molecular Weight:342.30 g/mol
- Merck Index:14,1961
- Smeltende Pt:234 °C (dec.)
- Opbevaringstemperatur:Stue temperatur
- EINECS:208-436-5
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



