Phthalsyre ≥99%
Glass bottle for solids, 250 g
Purity: 99% CAS No.: 88-99-3 Molecular Formula: C8H6O4 Formula Weight: 166.13 Flash Point: 168°(334°F) Odour: Odourless Beilstein Reference: 608199 Merck Reference: 14,7371 MDL No.: MFCD00002467
: Pathalic acid
- :88-99-3
- :C₈H₆O₄
- :MFCD00002467
- :166.13 g/mol
- :14,7371
- :315…317 °C (1013 hPa)
- :205 °C (dec.)
- :1,593
- :168 °C (334 °F)
- :Stue temperatur
- :201-873-2
Documents
- Pakning:Glass bottle for solids
- Pakn:250 g