Orders LinkContactUsLinkComponent
Phthalsyre ≥99%
Phthalsyre ≥99%
Glass bottle for solids, 250 g
  A14450.30
Phthalsyre ≥99%
  A14450.30
 :  88-99-3
Phthalsyre ≥99%
Phthalsyre ≥99%
Glass bottle for solids, 250 g
 :  88-99-3
471,00 kr
1 * 250 g
Purity: 99% CAS No.: 88-99-3 Molecular Formula: C8H6O4 Formula Weight: 166.13 Flash Point: 168°(334°F) Odour: Odourless Beilstein Reference: 608199 Merck Reference: 14,7371 MDL No.: MFCD00002467
 :  Pathalic acid
  •  :88-99-3
  •  :C₈H₆O₄
  •  :MFCD00002467
  •  :166.13 g/mol
  •  :14,7371
  •  :315…317 °C (1013 hPa)
  •  :205 °C (dec.)
  •  :1,593
  •  :168 °C (334 °F)
  •  :Stue temperatur
  •  :201-873-2
  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g