Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phthalsyre ≥99%

Phthalsyre ≥99%

Glass bottle for solids, 5.000 g
Katalog # A14450.0I
Irriterende
Leverandør:   Thermo Fisher Scientific
Phthalsyre ≥99%
Katalog # A14450.0I
CAS nummer:  88-99-3
Phthalsyre ≥99%
Phthalsyre ≥99%

Glass bottle for solids, 5.000 g

CAS nummer:  88-99-3
Price
4.040,00 kr
Log ind for tilgængelighed
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 88-99-3 Molecular Formula: C8H6O4 Formula Weight: 166.13 Flash Point: 168°(334°F) Odour: Odourless Beilstein Reference: 608199 Merck Reference: 14,7371 MDL No.: MFCD00002467

Synonymer:  Pathalic acid
  • CAS nummer:88-99-3
  • Formel:C₈H₆O₄
  • MDL nummer:MFCD00002467
  • Molecular Weight:166.13 g/mol
  • Merck Index:14,7371
  • Kogende Pt:315…317 °C (1013 hPa)
  • Smeltende Pt:205 °C (dec.)
  • Tæthed:1,593
  • Flash Pt:168 °C (334 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-873-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5.000 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.