Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Phthalsyre ≥99%

Glass bottle for solids, 1.000 g
Katalog # A14450.0B
Irriterende
Leverandør:   Thermo Fisher Scientific
Phthalsyre ≥99%
Katalog # A14450.0B
CAS nummer:  88-99-3
Phthalsyre ≥99%
Phthalsyre ≥99%

Glass bottle for solids, 1.000 g

CAS nummer:  88-99-3
Price
1.230,00 kr
Log ind for tilgængelighed
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 88-99-3 Molecular Formula: C8H6O4 Formula Weight: 166.13 Flash Point: 168°(334°F) Odour: Odourless Beilstein Reference: 608199 Merck Reference: 14,7371 MDL No.: MFCD00002467

Synonymer:  Pathalic acid
  • CAS nummer:88-99-3
  • Formel:C₈H₆O₄
  • MDL nummer:MFCD00002467
  • Molecular Weight:166.13 g/mol
  • Merck Index:14,7371
  • Kogende Pt:315…317 °C (1013 hPa)
  • Smeltende Pt:205 °C (dec.)
  • Tæthed:1,593
  • Flash Pt:168 °C (334 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-873-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifikationer