Fluorbenzen ≥99%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 462-06-6 Molecular Formula: C6H5F Formula Weight: 96.10 Flash Point: -15°(-5°F) Refractive Index: 1.4660 Odour: Aromatic Melting Point: -42° Boiling Point: 84-85° Beilstein Reference: 1236623 Merck Reference: 14,4170 UN No.: 2387 MDL No.: MFCD00000280
Synonyms: Phenylfluoride
- CAS Number:462-06-6
- Formula:C₆H₅F
- MDL Number:MFCD00000280
- Molecular Weight:96.1 g/mol
- Merck Index:14,4170
- Boiling Pt:84…85 °C
- Melting Pt:–42 °C
- Density:1,025
- Flash Pt:–15 °C (5 °F)
- Storage Temperature:Stue temperatur
- EINECS:207-321-7
- UN:2387
- ADR:3,II
Documents
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:250 g



